missing translation for 'onlineSavingsMsg'
Learn More

PPAR alpha/NR1C1 Antibody (2297B) [FITC], Novus Biologicals™

Product Code. 30733954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733954 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733954 Supplier Novus Biologicals Supplier No. FAB24781F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PPAR alpha/NR1C1 Monoclonal antibody specifically detects PPAR alpha/NR1C1 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR alpha/NR1C1
Applications Immunocytochemistry
Classification Monoclonal
Clone 2297B
Conjugate FITC
Formulation PBS
Gene Alias alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452
Host Species Rabbit
Immunogen E. coli-derived recombinant human PPAR alpha/NR1C1, Met1-Asn99, Accession # Q07869
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5465
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.