missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPAR alpha Monoclonal Antibody (3B6/PPAR)
GREENER_CHOICE

Product Code. 11520123
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11520123 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11520123 Supplier Invitrogen™ Supplier No. MA1822

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-822 detects peroxisome proliferator activated receptors (PPAR) alpha from human, mouse, and rat tissues. MA1-822 has been successfully used in Western blot, Gel Shift and immunoprecipitation procedures. By Western blot, this antibody detects a ~52 kDa protein which corresponds to PPAR alpha from mouse adipose tissue extract. This antibody detects some non-specific bands on NIH-3T3 cell lysates. The MA1-822 immunogen is purified recombinant PPAR alpha protein.

Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR's). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR's are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPAR's can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. PPAR, like several other nuclear hormone receptors, heterodimerizes with retinoid X receptor (RXR) alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR alpha
Applications ChIP Assay, Flow Cytometry, Gel Shift, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3B6/PPAR
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PPARA
Gene Accession No. P23204, P37230, Q07869
Gene Alias 4933429D07Rik; AW742785; hPPAR; I79_022392; MGC2237; MGC2452; Nr1c1; nuclear hormone receptor; nuclear receptor; nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor alpha; peroxisome proliferative activated receptor, alpha; peroxisome proliferative activated receptor, alpha isoform 1; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor alpha; peroxisome proliferator-activated nuclear receptor alpha variant 3; peroxisome proliferator-activated receptor alpha; PPAR; PPAR ALPHA; Ppara; PPARALPHA; PPAR-alpha; proliferator-activated receptor alpha; transcription factor belonging to the nuclear receptor superfamily
Gene Symbols PPARA
Host Species Mouse
Immunogen Purified recombinant PPAR alpha protein.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19013, 25747, 5465
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.