missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAPDC3 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | PPAPDC3 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PPAPDC3 Polyclonal specifically detects PPAPDC3 in Rat samples. It is validated for Western Blot.Specifications
| PPAPDC3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| C9orf67, chromosome 9 open reading frame 67, EC 3.1.3, EC 3.1.3.-, FLJ14662, KIAA0515, MGC12921, NET39, phosphatidic acid phosphatase type 2 domain containing 3, Phosphatidic acid phosphatase type 2 domain-containing protein 3, probable lipid phosphate phosphatase PPAPDC3 | |
| The immunogen is a synthetic peptide directed towards the N terminal region of rat PPAPDC3 (NP_001012349.1). Peptide sequence EFLSLNQPPKGTQEPRSSGRKASGPSTQPPPSSDGARERRQSQQLPEEDC | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 84814 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title