missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPAPDC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12839012
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12839012 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12839012 Supplier Invitrogen™ Supplier No. PA520865

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Raji cell lysate. PA5-20865 can be used with blocking peptide PEP-0979.

PPAPDC2 is a phosphatase that dephosphorylates Presqualene diphosphate (PSDP) into presqualene monophosphate (PSMP), suggesting that it may have important role in innate immunity. PSDP is a bioactive lipid that rapidly remodels to PSMP upon cell activation. PPAPDC2 displays diphosphate phosphatase activity with a substrate preference for PSDP > FDP > phosphatidic acid. PPAPDC2 activity is independent of Mg2+ and has been identified in activated human neutrophil (PMN) extracts. It is widely expressed in human tissues. Recent studies shows PPAPDC2 is a functional isoprenoid diphosphate phosphatase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAPDC2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PLPP6
Gene Accession No. Q66H88, Q8IY26, Q9D4F2
Gene Alias 4932443D16Rik; bA6J24.6; FLJ46512; FLJ90191; MGC15483; PDP1; phosphatidic acid phosphatase type 2 domain containing 2; phosphatidic acid phosphatase type 2 domain-containing protein 2; Phospholipid phosphatase 6; PLPP6; polyisoprenoid diphosphate phosphatase type 1; PPAP2 domain-containing protein 2; Ppapdc2; Presqualene diphosphate phosphatase; PSDP; RP11-6J24.2
Gene Symbols PLPP6
Host Species Rabbit
Immunogen A 13 amino acid peptide from near the carboxy terminus human PPAPDC2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403313, 619549, 74411
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.