missing translation for 'onlineSavingsMsg'
Learn More

PPAPDC1B Polyclonal Antibody

Product Code. 12809032
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809032 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809032 Supplier Invitrogen Supplier No. PA520881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PPAPDC1B Polyclonal Antibody

PPAPDC1B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAPDC1B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PLPP5
Gene Accession No. Q3UMZ3, Q8NEB5
Gene Alias 1810019D05Rik; 2310022A04Rik; AW045471; diacylglycerol pyrophosphate like 1; diacylglycerol pyrophosphate phosphatase-like 1; DPPL1; HTPAP; phosphatidate phosphatase PPAPDC1B; phosphatidic acid phosphatase type 2 domain containing 1; phosphatidic acid phosphatase type 2 domain containing 1B; phosphatidic acid phosphatase type 2 domain-containing protein 1B; Phospholipid phosphatase 5; Plpp5; Ppapdc1; Ppapdc1b; testicular secretory protein Li 38
Gene Symbols PLPP5
Host Species Rabbit
Immunogen A 14 amino acid peptide near the amino terminus of human PPAPDC1B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71910, 84513
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.