missing translation for 'onlineSavingsMsg'
Learn More

PPAPDC1B/DPPL1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18424893 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424893 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424893 Supplier Novus Biologicals Supplier No. NBP176244AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPAPDC1B/DPPL1 Polyclonal antibody specifically detects PPAPDC1B/DPPL1 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAPDC1B/DPPL1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias diacylglycerol pyrophosphate like 1, DPPL1, EC 3.1.3.4, HTPAPphosphatidate phosphatase PPAPDC1B, phosphatidic acid phosphatase type 2 domain containing 1B, Phosphatidic acid phosphatase type 2 domain-containing protein 1B
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human PPAPDC1B. The immunogen is located within amino acids 30 - 80 of PPAPDC1B. Amino Acid Squence: LYRNPYVEAEYFPT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84513
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.