missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPAP2C Polyclonal antibody specifically detects PPAP2C in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PPAP2C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 10 - 15 ug/ml |
| Formulation | PBS |
| Gene Alias | EC 3.1.3.4, LPP2PAP2-g, PAP2c, PAP-2clipid phosphate phosphohydrolase 2, PAP2-G, PAP2-gamma, Phosphatidate phosphohydrolase type 2c, Phosphatidic acid phosphatase 2c, phosphatidic acid phosphatase type 2C, phosphatidic acid phosphohydrolase type 2c, type-2 phosphatidic acid phosphatase-gamma |
| Host Species | Rabbit |
| Immunogen | Unique peptide derived from the human lysophospholipid phosphatase 2 protein. (Uniprot: O43688) |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?