missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAP2C Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11903-100UG
This item is not returnable.
View return policy
Description
PPAP2C Polyclonal antibody specifically detects PPAP2C in Human samples. It is validated for Western Blot
Specifications
| PPAP2C | |
| Polyclonal | |
| Western Blot 10 - 15 ug/ml | |
| EC 3.1.3.4, LPP2PAP2-g, PAP2c, PAP-2clipid phosphate phosphohydrolase 2, PAP2-G, PAP2-gamma, Phosphatidate phosphohydrolase type 2c, Phosphatidic acid phosphatase 2c, phosphatidic acid phosphatase type 2C, phosphatidic acid phosphohydrolase type 2c, type-2 phosphatidic acid phosphatase-gamma | |
| Unique peptide derived from the human lysophospholipid phosphatase 2 protein. (Uniprot: O43688) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8612 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction