missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPAP2C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30860-100ug
This item is not returnable.
View return policy
Description
PPAP2C Polyclonal antibody specifically detects PPAP2C in Human,Porcine samples. It is validated for ELISA,Western Blot
Specifications
| PPAP2C | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8612 | |
| Store at -70°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| EC 3.1.3.4, LPP2PAP2-g, PAP2c, PAP-2clipid phosphate phosphohydrolase 2, PAP2-G, PAP2-gamma, Phosphatidate phosphohydrolase type 2c, Phosphatidic acid phosphatase 2c, phosphatidic acid phosphatase type 2C, phosphatidic acid phosphohydrolase type 2c, type-2 phosphatidic acid phosphatase-gamma | |
| Synthetic peptide derived from the PPAP2C | |
| 100 μg | |
| Primary | |
| Human, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction