missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPAP2B Recombinant Rabbit Monoclonal Antibody (6E10N3)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558135

 View more versions of this product

Product Code. 30283972

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MQNYKYDKAI VPESKNGGSP ALNNNPRRSG SKRVLLICLD LFCLFMAGLP FLIIETSTIK PYHRGFYCND ESIKYPLKTG ETINDAVLCA VGIVIAILAI.

Human vascular endothelial growth factor produced in E.Coli is a double, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 38231 Dalton. The rHuVEGF is purified by proprietary chromatographic techniques.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPAP2B
Recombinant Monoclonal
1.81 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
O14495, P97544, Q99JY8
PLPP3
Synthetic peptide.
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
6E10N3
Unconjugated
PLPP3
1110003O22Rik; 2610002D05Rik; AV025606; D4Bwg0538e; D4Bwg1535e; differentially expressed in rat intestine 42; Dri42; EC 3.1.3.4; ER transmembrane protein Dri 42; lipid phosphate phosphohydrolase 3; Lpp3; MGC15306; PAP2 beta; PAP2B; PAP-2b; PAP2-b; PAP2-beta; phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; phospholipid phosphatase 3; PLPP3; Ppab2b; PPAP2B; type-2 phosphatidic acid phosphatase-beta; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP
Rabbit
Protein A
RUO
192270, 67916, 8613
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.