missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPAP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA112730
This item is not returnable.
View return policy
Description
A suggested positive control for this product is human brain lysate.
Human vascular endothelial growth factor produced in E.Coli is a double, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 38231 Dalton. The rHuVEGF is purified by proprietary chromatographic techniques.
Specifications
| PPAP2B | |
| Polyclonal | |
| PBS with 0.08% sodium azide | |
| O14495, P97544, Q99JY8 | |
| PLPP3 | |
| Synthetic peptide corresponding to phosphatidic acid phosphatase 2B. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PLPP3 | |
| 1110003O22Rik; 2610002D05Rik; AV025606; D4Bwg0538e; D4Bwg1535e; differentially expressed in rat intestine 42; Dri42; EC 3.1.3.4; ER transmembrane protein Dri 42; lipid phosphate phosphohydrolase 3; Lpp3; MGC15306; PAP2 beta; PAP2B; PAP-2b; PAP2-b; PAP2-beta; phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; phospholipid phosphatase 3; PLPP3; Ppab2b; PPAP2B; type-2 phosphatidic acid phosphatase-beta; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192270, 67916, 8613 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction