missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPAP2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30861-25ug
This item is not returnable.
View return policy
Description
PPAP2B Polyclonal antibody specifically detects PPAP2B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifikationer
| PPAP2B | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| Dri42, EC 3.1.3.4, lipid phosphate phosphohydrolase 3, LPP3PAP2 beta, MGC15306, PAP-2b, PAP2B, PAP2-beta, Phosphatidate phosphohydrolase type 2b, Phosphatidic acid phosphatase 2b, phosphatidic acid phosphatase type 2B, type-2 phosphatidic acid phosphatase-beta, vascular endothelial growth factor and type I collagen inducible, Vascular endothelial growth factor and type I collagen-inducible protein, VCIP | |
| Synthetic peptide derived from the PPAP2B | |
| 25 μg | |
| Protein Phosphatase | |
| 8613 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering