missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PPAP2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£162.00 - £386.00

Specifications

Antigen PPAP2B
Dilution Western Blot : 10-15 μg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067063
View Documents
Novus Biologicals™
NBP3-30837-25ug
25 μg
£162.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30066811
View Documents
Novus Biologicals™
NBP3-30837-100ug
100 μg
£386.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PPAP2B Polyclonal antibody specifically detects PPAP2B in Human samples. It is validated for Western Blot
Specifications

Specifications

PPAP2B
Western Blot
Unconjugated
Rabbit
Protein Phosphatase
Phosphate buffered saline
8613
IgG
Ammonium sulfate precipitation
Western Blot : 10-15 μg/mL
Polyclonal
Purified
RUO
Human
Dri42, EC 3.1.3.4, lipid phosphate phosphohydrolase 3, LPP3PAP2 beta, MGC15306, PAP-2b, PAP2B, PAP2-beta, Phosphatidate phosphohydrolase type 2b, Phosphatidic acid phosphatase 2b, phosphatidic acid phosphatase type 2B, type-2 phosphatidic acid phosphatase-beta, vascular endothelial growth factor and type I collagen inducible, Vascular endothelial growth factor and type I collagen-inducible protein, VCIP
Rabbits were immunized with a unique peptide derived from the human PPAP2B protein.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.