missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPAP2A Polyclonal Antibody

Description
Peptide sequence: QIYPFQRGFF CKDNSINYPY HDSTVTSTVL ILVGVGLPIS SIILGETLSV Sequence homology: Guinea Pig: 86%; Human: 100%; Mouse: 79%; Rat: 93%.
Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.
Specifications
Specifications
| Antigen | PPAP2A |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PLPP1 |
| Gene Accession No. | O14494 |
| Gene Alias | 35 kDa PAP; Hic53; Hpic53; hydrogen peroxide inducible protein 53; Hydrogen peroxide-inducible protein 53; Lipid phosphate phosphohydrolase 1; lipid phosphate phosphohydrolase 1a; LLP1a; Lpp1; LPP-1; magnesium-independent type 2 phosphatidic acid phosphatase; mPAP; PAP2; PAP2A; PAP-2a; PAP2a2; PAP2-alpha; PAP2alpha2; PAPalpha1; phosphatidate phosphohydrolase type 2a; Phosphatidic acid phosphatase 2a; phosphatidic acid phosphatase 2a2; phosphatidic acid phosphatase type 2A; phosphatidic acid phosphohydrolase type 2a; phospholipid phosphatase 1; Plpp1; Ppap2a; PPAP2A; LPP1; type-2 phosphatidic acid phosphatase alpha; Unknown (protein for MGC:133873); WUN |
| Gene Symbols | PLPP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?