missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPAP2A Monoclonal Antibody (OTI1H4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526283
This item is not returnable.
View return policy
Description
PPAP2A Monoclonal Antibody for Western Blot, IHC (P)
Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.
Specifications
| PPAP2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14494 | |
| PLPP1 | |
| Full length human recombinant protein of PPAP2A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H4 | |
| Unconjugated | |
| PLPP1 | |
| 35 kDa PAP; Hic53; Hpic53; hydrogen peroxide inducible protein 53; Hydrogen peroxide-inducible protein 53; Lipid phosphate phosphohydrolase 1; lipid phosphate phosphohydrolase 1a; LLP1a; Lpp1; LPP-1; magnesium-independent type 2 phosphatidic acid phosphatase; mPAP; PAP2; PAP2A; PAP-2a; PAP2a2; PAP2-alpha; PAP2alpha2; PAPalpha1; phosphatidate phosphohydrolase type 2a; Phosphatidic acid phosphatase 2a; phosphatidic acid phosphatase 2a2; phosphatidic acid phosphatase type 2A; phosphatidic acid phosphohydrolase type 2a; phospholipid phosphatase 1; Plpp1; Ppap2a; PPAP2A; LPP1; type-2 phosphatidic acid phosphatase alpha; Unknown (protein for MGC:133873); WUN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8611 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction