missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAP2A Antibody (OTI1H4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73564F
This item is not returnable.
View return policy
Description
PPAP2A Monoclonal specifically detects PPAP2A in Human samples. It is validated for Immunohistochemistry.
Specifications
| PPAP2A | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PPAP2A | |
| Full length recombinant protein of human PPAP2A (NP_003702) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| OTI1H4 | |
| Immunohistochemistry | |
| EC 3.1.3.4, lipid phosphate phosphohydrolase 1, lipid phosphate phosphohydrolase 1a, LLP1a, LPP1PAP2a2, PAP2, PAP2a, PAP2-alpha, PAP-2aPAP2alpha2, PAPalpha1, Phosphatidate phosphohydrolase type 2a, Phosphatidic acid phosphatase 2a, phosphatidic acid phosphatase type 2A, phosphatidic acid phosphohydrolase type 2a, type 2 phosphatidic acid phosphohydrolase, type-2 phosphatidic acid phosphatase alpha | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 8611.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction