missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPAP2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30836-25ug
This item is not returnable.
View return policy
Description
PPAP2A Polyclonal antibody specifically detects PPAP2A in Human samples. It is validated for Western Blot
Specifications
| PPAP2A | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| EC 3.1.3.4, lipid phosphate phosphohydrolase 1, lipid phosphate phosphohydrolase 1a, LLP1a, LPP1PAP2a2, PAP2, PAP2a, PAP2-alpha, PAP-2aPAP2alpha2, PAPalpha1, Phosphatidate phosphohydrolase type 2a, Phosphatidic acid phosphatase 2a, phosphatidic acid phosphatase type 2A, phosphatidic acid phosphohydrolase type 2a, type 2 phosphatidic acid phosphohydrolase, type-2 phosphatidic acid phosphatase alpha | |
| Synthetic peptide derived from the PPAP2A protein | |
| 25 μg | |
| Cell Cycle and Replication | |
| 8611 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion