missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPAP2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30836-100ug
This item is not returnable.
View return policy
Description
PPAP2A Polyclonal antibody specifically detects PPAP2A in Human samples. It is validated for Western Blot
Specifications
| PPAP2A | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| EC 3.1.3.4, lipid phosphate phosphohydrolase 1, lipid phosphate phosphohydrolase 1a, LLP1a, LPP1PAP2a2, PAP2, PAP2a, PAP2-alpha, PAP-2aPAP2alpha2, PAPalpha1, Phosphatidate phosphohydrolase type 2a, Phosphatidic acid phosphatase 2a, phosphatidic acid phosphatase type 2A, phosphatidic acid phosphohydrolase type 2a, type 2 phosphatidic acid phosphohydrolase, type-2 phosphatidic acid phosphatase alpha | |
| Synthetic peptide derived from the PPAP2A protein | |
| 100 μg | |
| Cell Cycle and Replication | |
| 8611 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction