missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PPADS tetrasodium salt

Description
PPADS tetrasodium salt is a non-selective P2 purinergic antagonist. Blocks recombinant P2X1, P2X2, P2X3, P2X5 (IC50 = 1 - 2.6 μM), native P2Y2-like (IC50 ∼ 0.9 mM), and recombinant P2Y4 (IC50 ∼ 15 mM) receptors. PPADS tetrasodium salt delays onset of calcium responses to mild hypoosmotic stress in cortical slices. PPADS tetrasodium salt blocks P2X7 and reduces clinical and histological graft-versus-host disease development in a humanised mouse model.
2'5'-isomer also available.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 599.3 |
| Purity | >98% |
| Molecular Weight (g/mol) | 599.296 |
| Chemical Name or Material | PPADS tetrasodium salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?