Learn More
PPA2 Mouse, anti-Human, Clone: 1C2E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68468-1-IG-20UL
This item is not returnable.
View return policy
Description
The PPA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPA2. This antibody recognizes human, mouse, rat antigen. The PPA2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PPA2 | |
| 1C2E3 | |
| FLJ20459, HSPC124, PPA2, PPase 2, pyrophosphatase (inorganic) 2, Pyrophosphatase SID6 306, SID6 306 | |
| 20 μL | |
| 27068 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG1 |