missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPA2 Antibody (OTI7C1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73563APC
This item is not returnable.
View return policy
Description
PPA2 Monoclonal antibody specifically detects PPA2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PPA2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI7C1 | |
| Western Blot, Immunohistochemistry | |
| EC 3.6.1.1, FLJ20459, MGC49850, mitochondrial, pyrophosphatase (inorganic) 2 | |
| Full length human recombinant protein of human PPA2(NP_008834) produced in E.coli. | |
| 0.1 mL | |
| Cancer | |
| 27068 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction