missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPA1 Recombinant Rabbit Monoclonal Antibody (24GB1365)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | PPA1 |
|---|---|
| Clone | 24GB1365 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275393
![]() |
Invitrogen™
MA557430 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274993
|
Invitrogen™
MA557431 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPA1 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.Specifications
| PPA1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Ppa1 | |
| 2010317E03Rik; cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; HEL-S-66p; inorganic diphosphatase; inorganic diphosphatase 1; inorganic pyrophosphatase; inorganic pyrophosphatase 1; IOPPP; Pp; PP1; Ppa1; PPase; Pyp; pyrophosphatase (inorganic) 1; pyrophosphatase 1; pyrophosphate phospho-hydrolase; RP11-367H5.1; SID6-8061 | |
| Ppa1 | |
| Primary | |
| Affinity chromatography |
| 24GB1365 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15181, Q9D819 | |
| 294504, 5464, 67895 | |
| A synthesized peptide derived from human Pyrophosphatase 1 (aa 1-45). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title