missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPA1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA554105

Product Code. 15732041

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GFSTEERAAP FSLEYRVFLK NEKGQYISPF HDIPIYADKD VFHMVVEVPR WSNAK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 91%.

The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPA1
Polyclonal
Unconjugated
Ppa1
2010317E03Rik; cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; HEL-S-66p; inorganic diphosphatase; inorganic diphosphatase 1; inorganic pyrophosphatase; inorganic pyrophosphatase 1; IOPPP; Pp; PP1; Ppa1; PPase; Pyp; pyrophosphatase (inorganic) 1; pyrophosphatase 1; pyrophosphate phospho-hydrolase; RP11-367H5.1; SID6-8061
Rabbit
Antigen affinity chromatography
RUO
5464
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q15181
Ppa1
Recombinant protein corresponding to Human PPA1. Recombinant protein control fragment (Product #RP-92488).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.