missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PP5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16332225
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16332225 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16332225 Supplier Invitrogen™ Supplier No. PA585283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (85%), Xenopus laevis (85%), Rhesus Monkey (99%), Bovine (97%). Positive Control: 293T, A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP5
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ppp5c
Gene Accession No. P53041
Gene Alias ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5
Gene Symbols Ppp5c
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP5C. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5536
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.