missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP5 Monoclonal Antibody (12F7), FITC

Description
1 μg/mL of MA5-45365 was sufficient for detection of PP5 in 15 μg of human A431 lysates by ECL analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 58kDa.
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | PP5 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 12F7 |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 |
| Gene | Ppp5c |
| Gene Accession No. | P53041 |
| Gene Alias | ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?