missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PP5 Polyclonal antibody specifically detects PP5 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PP5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 5-10 μg/mL |
| Formulation | Phosphate buffered saline |
| Gene Alias | EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5 |
| Host Species | Rabbit |
| Immunogen | Rabbits were immunized with a Synthetic peptide derived from the human PP5 |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?