missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP5 Antibody (2E11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59307F
This item is not returnable.
View return policy
Description
PP5 Monoclonal specifically detects PP5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| PP5 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PPP5C | |
| Full length human PP5 protein | |
| 100 μL | |
| Primary | |
| Detects 58kDa. | |
| Store at 4C in the dark. | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence | |
| 200000000000 | |
| Immunocytochemistry/Immunofluorescence | |
| EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5 | |
| Mouse | |
| Protein G purified | |
| Cell Cycle and Replication | |
| 5536 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction