missing translation for 'onlineSavingsMsg'
Learn More

PP2D1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £513.00

Specifications

Antigen PP2D1
Dilution Immunohistochemistry-Paraffin 1:2500 - 1:5000
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18250724
View Documents
Novus Biologicals
NBP2-57748
100 μL
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18614717
View Documents
Novus Biologicals
NBP2-57748-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PP2D1 Polyclonal specifically detects PP2D1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PP2D1
Immunohistochemistry (Paraffin)
Unconjugated
RUO
C3orf48, Chromosome 3 Open Reading Frame 48, Protein Phosphatase 2C-Like Domain Containing 1, Protein Phosphatase 2C-Like Domain-Containing Protein 1
PP2D1
IgG
Affinity Purified
Immunohistochemistry-Paraffin 1:2500 - 1:5000
Polyclonal
Rabbit
Human
151649
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PSYQMTTDEQQIINSFYTVFREEYAAIEDLFSAINKTEAVRCEYEDTHKAFAKAFWRMDRLLGLGRKEVSRVQWSGC
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.