missing translation for 'onlineSavingsMsg'
Learn More

PP2C gamma/PPM1G Antibody (112), FITC, Novus Biologicals™

Product Code. 18350671 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350671 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350671 Supplier Novus Biologicals Supplier No. NBP289858F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PP2C gamma/PPM1G Monoclonal specifically detects PP2C gamma/PPM1G in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2C gamma/PPM1G
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 112
Conjugate FITC
Formulation PBS
Gene Alias EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2C-gamma, PP2CGAMMA, PPM1C, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PP2C gamma/PPM1G (O15355; Met317-Asp546).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5496
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.