missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2C gamma/PPM1G Antibody (001), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
PP2C gamma/PPM1G Monoclonal antibody specifically detects PP2C gamma/PPM1G in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2C-gamma, PP2CGAMMA, PPM1C, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G |
| Host Species | Rabbit |
| Purification Method | Protein A purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?