missing translation for 'onlineSavingsMsg'
Learn More

PP2C beta/PPM1B Antibody (OTI3C9), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18423127 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423127 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423127 Supplier Novus Biologicals Supplier No. NBP273548AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PP2C beta/PPM1B Monoclonal antibody specifically detects PP2C beta/PPM1B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2C beta/PPM1B
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone OTI3C9
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.16, MGC21657, PP2C-beta, PP2C-beta-X, PP2CBmagnesium-dependent, beta isoform, protein phosphatase 1B, Protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein, protein phosphatase, Mg2+/Mn2+ dependent, 1B
Host Species Mouse
Immunogen Full length recombinant protein of human PP2C beta/PPM1B (NP_002697) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5495
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.