missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2C alpha/PPM1A Antibody (p6c7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04333CL1
This item is not returnable.
View return policy
Description
PP2C alpha/PPM1A Monoclonal antibody specifically detects PP2C alpha/PPM1A in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| PP2C alpha/PPM1A | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
| Recombinant human PP2C alpha/PPM1A (1-382aa) purified from E. coli (NP_066283). | |
| 0.1 mL | |
| Signal Transduction | |
| 5494 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| p6c7 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction