missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PP2C alpha/PPM1A Polyclonal antibody specifically detects PP2C alpha/PPM1A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PP2C alpha/PPM1A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human PP2C alpha/PPM1A fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?