missing translation for 'onlineSavingsMsg'
Learn More

PP2C alpha/PPM1A Antibody, DyLight 550, Novus Biologicals™

Product Code. 18373517 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373517 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373517 Supplier Novus Biologicals Supplier No. NBP300058R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PP2C alpha/PPM1A Polyclonal specifically detects PP2C alpha/PPM1A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2C alpha/PPM1A
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PP2C alpha/PPM1A (P35813-1; Met1-Trp382).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5494
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.