missing translation for 'onlineSavingsMsg'
Learn More

PP2C alpha/PPM1A Antibody (498922) [PE], Novus Biologicals™

Product Code. 30735297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735297 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735297 Supplier Novus Biologicals Supplier No. FAB4150P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PP2C alpha/PPM1A Monoclonal antibody specifically detects PP2C alpha/PPM1A in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2C alpha/PPM1A
Applications Western Blot
Classification Monoclonal
Clone 498922
Conjugate PE
Formulation PBS
Gene Alias EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A
Host Species Mouse
Immunogen E. coli-derived recombinant human PP2C alpha/PPM1A, Met1-Gly324, Accession # P35813
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5494
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.