missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2A alpha, p Tyr307 Antibody (SR1588), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21512-50ul
This item is not returnable.
View return policy
Description
PP2A alpha Monoclonal antibody specifically detects PP2A alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| PP2A alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1588 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
| A synthesized peptide derived from human phospho-PP2A alpha [Tyr307] (Uniprot #: P67775) | |
| 50 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Phospho Specific, Protein Phosphatase, Signal Transduction | |
| 5515 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction