missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PP2A alpha (Tyr307) Recombinant Rabbit Monoclonal Antibody (9B7Z9)
GREENER_CHOICE

Product Code. 17156533
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17156533 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17156533 Supplier Invitrogen™ Supplier No. MA535878

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: PDYFL.

In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. An additional protein phosphatase catalytic subunit, PPX (also known as PP4) is a putative member of a novel PP family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PP2A alpha (Tyr307)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9B7Z9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP2CA
Gene Accession No. P63330, P67775
Gene Alias B55-GAMMA; hypothetical protein; IMYPNO; IMYPNO1; Methyl-PPP2CA; MGC33570; PP2A; PP2A alpha; PP2A C terminal; PP2A L9; PP2A Leucine 9; Pp2a1; PP2A-alpha; PP2Ac; PP2CA; PP2Calpha; Ppp2ca; PR52; PR55G; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A; protein phosphatase-2A-alpha; R75353; replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; unnamed protein product
Gene Symbols PPP2CA
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around Y307 of human PP2A Catalytic alpha (P67775).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19052, 5515
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.