missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PP2A alpha/beta Antibody, Novus Biologicals™

Sheep Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30778-25ug

 View more versions of this product

Product Code. 30066798

  • £162.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PP2A alpha/beta Polyclonal antibody specifically detects PP2A alpha/beta in Bovine,Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PP2A alpha/beta
Polyclonal
Western Blot : 1:500-1:1000
PPP2CA, PPP2CB, Replication protein C, RP-C, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit beta isoform
A peptide conjugated to KLH
25 μg
Primary
Bovine, Human, Mouse, Rat
Purified
Western Blot
Unconjugated
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
5515
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis