missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PP2A alpha Monoclonal antibody specifically detects PP2A alpha in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PP2A alpha |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | H5D12 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform |
| Host Species | Mouse |
| Immunogen | GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?