missing translation for 'onlineSavingsMsg'
Learn More

PP2, MedChemExpress

Product Code. 16478517
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478517

Brand: Medchem Express HY13805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PP2 is a reversible and ATP-competitive Src family kinases inhibitor with IC50s of 4 and 5 nM for Lck and Fyn, respectively.

PP2 is a reversible and ATP-competitive Src family kinases inhibitor with IC50s of 4 and 5 nM for Lck and Fyn, respectively.

Specifications

Chemical Name or Material PP2
Physical Form Solid
CAS 172889-27-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H16ClN5
For Use With (Application) Cancer-Kinase/protease
Synonym AGL 1879
Solubility Information DMSO : 50 mg/mL (165.69 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC1=C2C(N(C(C)(C)C)N=C2C3=CC=C(Cl)C=C3)=NC=N1
Molecular Weight (g/mol) 301.77
Formula Weight 301.77
Percent Purity 98.96%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.