missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP14/Glycodelin Mouse anti-Human, Clone: 16, Novus Biologicals™
Mouse Monoclonal Antibody
£332.00
Specifications
| Antigen | PP14/Glycodelin |
|---|---|
| Clone | 16 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Applications | ELISA |
Description
PP14/Glycodelin Monoclonal specifically detects PP14/Glycodelin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PP14/Glycodelin | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein | |
| Placental protein 14 (glycodelin A) purified from second trimester amniotic fluid | |
| Primary | |
| Store at 4°C. Do not freeze. |
| 16 | |
| ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 5047 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title