missing translation for 'onlineSavingsMsg'
Learn More

PP14/Glycodelin Antibody (16), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18401945 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401945 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401945 Supplier Novus Biologicals Supplier No. NBP105115AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PP14/Glycodelin Monoclonal antibody specifically detects PP14/Glycodelin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP14/Glycodelin
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 16
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein
Host Species Mouse
Immunogen Placental protein 14 (glycodelin A) purified from second trimester amniotic fluid
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 5047
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.