missing translation for 'onlineSavingsMsg'
Learn More

PP121, MedChemExpress

Product Code. p-200097015
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16494936

Brand: Medchem Express HY10372

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PP121 is a multi-targeted kinase inhibitor with IC50s of 10, 60, 12, 14, 2 nM for mTOR, DNK-PK, VEGFR2, Src, PDGFR, respectively.

PP121 is a multi-targeted kinase inhibitor with IC50s of 10, 60, 12, 14, 2 nM for mTOR, DNK-PK, VEGFR2, Src, PDGFR, respectively.

Specifications

Chemical Name or Material PP121
Physical Form Solid
CAS 1092788-83-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H17N7
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (62.63 mM; Need ultrasonic)
SMILES NC1=C2C(C3=CC4=C(N=C3)NC=C4)=NN(C2=NC=N1)C5CCCC5
Molecular Weight (g/mol) 319.36
Formula Weight 319.36
Percent Purity 99.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.