missing translation for 'onlineSavingsMsg'
Learn More

PP121, MedChemExpress

Product Code. 16494936
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494936 100 mg 100mg
16460267 50 mg 50mg
16443908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494936 Supplier Medchem Express Supplier No. HY10372
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PP121 is a multi-targeted kinase inhibitor with IC50s of 10, 60, 12, 14, 2 nM for mTOR, DNK-PK, VEGFR2, Src, PDGFR, respectively.

PP121 is a multi-targeted kinase inhibitor with IC50s of 10, 60, 12, 14, 2 nM for mTOR, DNK-PK, VEGFR2, Src, PDGFR, respectively.

Specifications

Chemical Name or Material PP121
Physical Form Solid
CAS 1092788-83-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H17N7
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (62.63 mM; Need ultrasonic)
SMILES NC1=C2C(C3=CC4=C(N=C3)NC=C4)=NN(C2=NC=N1)C5CCCC5
Molecular Weight (g/mol) 319.36
Formula Weight 319.36
Percent Purity 99.08%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.