Learn More
Invitrogen™ PP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143968
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue. IHC: human lung cancer tissue, human placenta tissue. ICC/IF: Caco-2 cell. Flow: U87 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.
Specifications
| PP11 | |
| Polyclonal | |
| Unconjugated | |
| ENDOU | |
| 22 serine protease; 26 serine protease; endonuclease, poly(U) specific; endonuclease, poly(U)-specific; endonuclease, polyU-specific; ENDOU; P11; placental protein 11; placental protein 11 related; placental protein 11-related protein; Poly(U)-specific endoribonuclease; PP11; Pp11r; PP11-related protein; Protein endoU; PRSS26; T-cell-specific protein 30; Tcl-30; Uridylate-specific endoribonuclease | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8909 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P21128 | |
| ENDOU | |
| E.coli-derived human PP11/ENDOU recombinant protein (Position: K20-T386). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.