Learn More
Eurofins DiscoverX PP1α Protein
Description
PP1α Protein is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Increased PP1 activity has been observed in the end stage of heart failure. Studies in both human and mice suggest that PP1 is an important regulator of cardiac function. Mouse studies also suggest that PP1 functions as a suppressor of learning and memory. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.
GenBank NM_002708. The PP1alpha protein sequence is identical to the human PP1 alpha described inGenBank NM_002708. The DNA sequence, however, is more closely related to the rabbit cDNA described inGenBank X14832.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Hepes/NaOH pH 7.5, 300 mM NaCl, 50% glycerol, 0.1 mM EGTA, 1 mM MnCl2, 0.03% Brij-35, 0.1 mM PMSF, 1 mM benzamidine, 0.1% 2-mercaptoethanol. Liquid at -20°C. |
| Molecular Weight (g/mol) | 37.6 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | Untagged |
| Expression System | Recombinant enzyme expressed in E. coli cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥59% |
| Protein | PP1α |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.