missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP1 Inhibitor-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | PP1 Inhibitor-2 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643159
|
Novus Biologicals
NBP2-48801-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661696
|
Novus Biologicals
NBP2-48801 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP1 Inhibitor-2 Polyclonal antibody specifically detects PP1 Inhibitor-2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| PP1 Inhibitor-2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.2), 40% Glycerol | |
| 5504 | |
| IgG | |
| Immunogen affinity purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| IPP-2, IPP2phosphatase inhibitor 2, MGC87148, phosphoprotein phosphatase, protein phosphatase 1, regulatory (inhibitor) subunit 2, protein phosphatase inhibitor 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SQKWEEMNILATYHPADKDYGLMK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title