missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP1 beta Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA112380
This item is not returnable.
View return policy
Description
PP1 beta Polyclonal Antibody for Western Blot, IP
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| PP1 beta | |
| Polyclonal | |
| PBS with 0.08% sodium azide | |
| P62140, P62141, P62142, Q3SWW9 | |
| Ppp1cb | |
| Synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Ppp1cb | |
| 1200010B19; epididymis secretory sperm binding protein Li 80p; HEL-S-80p; hypothetical protein LOC538829; MGC21657; PP1B; PP-1B; PP1beta; PP1-BETA; PP2CB; PP2CBETA; PP2C-beta-X; PPC2BETAX; PPP1CB; PPP1CD; protein phosphatase 1; protein phosphatase 1 catalytic subunit beta; protein phosphatase 1, catalytic subunit, beta isoform; protein phosphatase 1, catalytic subunit, beta isozyme; protein phosphatase 1, catalytic subunit, delta isoform; protein phosphatase 1-beta; protein phosphatase 1-delta; protein phosphatase-1a; serine/threonine protein phosphatase PP1-beta catalytic subunit; serine/threonine-protein phosphatase PP1-beta catalytic subunit | |
| Sheep | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 19046, 25594, 538829, 5500 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction