missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP1 beta Recombinant Rabbit Monoclonal Antibody (2E9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532936
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| PP1 beta | |
| Recombinant Monoclonal | |
| 0.3 mg/mL | |
| 50mM tris glycine with 0.05% BSA, 40% glycerol, 0.15M NaCl and 0.01% sodium azide; pH 7.4 | |
| P62140, P62141, P62142 | |
| Ppp1cb | |
| Recombinant full length Human PPP1CB. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2E9 | |
| Unconjugated | |
| Ppp1cb | |
| 1200010B19; epididymis secretory sperm binding protein Li 80p; HEL-S-80p; hypothetical protein LOC538829; MGC21657; PP1B; PP-1B; PP1beta; PP1-BETA; PP2CB; PP2CBETA; PP2C-beta-X; PPC2BETAX; PPP1CB; PPP1CD; protein phosphatase 1; protein phosphatase 1 catalytic subunit beta; protein phosphatase 1, catalytic subunit, beta isoform; protein phosphatase 1, catalytic subunit, beta isozyme; protein phosphatase 1, catalytic subunit, delta isoform; protein phosphatase 1-beta; protein phosphatase 1-delta; protein phosphatase-1a; serine/threonine protein phosphatase PP1-beta catalytic subunit; serine/threonine-protein phosphatase PP1-beta catalytic subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19046, 25594, 5500 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction