missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PP1 alpha/PPP1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | PP1 alpha/PPP1A |
|---|---|
| Dilution | Western Blot : 1:500-1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067000
|
Novus Biologicals™
NBP3-30764-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066867
|
Novus Biologicals™
NBP3-30764-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP1 alpha/PPP1A Polyclonal antibody specifically detects PP1 alpha/PPP1A in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| PP1 alpha/PPP1A | |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Phosphatase | |
| Phosphate buffered saline | |
| 5499 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1:500-1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
| The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Arg-Pro-Ile-Thr-Pro-Pro-Arg-Asn-Ser-AlaLys-Ala-Lys-Lys-COOH. This peptide antibody corresponds to C -terminal peptide of PP1 alpha/PPP1A catalytic subunit having a MW of 38kDa. The sequence used is amino acid 317-330. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title